Order Entry
United States
ContactUsLinkComponent
 
Anti-OR4F17 Rabbit Polyclonal Antibody
  89519-886
 :  Abgent
Anti-OR4F17 Rabbit Polyclonal Antibody
  89519-886
 :  Abgent
 :  AP16349A

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Olfactory Receptor, Family 4, Subfamily F Member 17
  • Antigen Symbol:
    OR4F17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    N-Terminal
  • Gene ID:
    81099
  • Antigen Synonyms:
    OR4F11P|OR4F18|OR4F19
  • Amino Acid Number:
    61 to 89
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    34.212 kDa
  • Sequence:
    TDFFSQRKVISFKGC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.39mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This OR4F17 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 61-89 amino acids from the N-terminal region of human OR4F17.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89519-886
  • Supplier no.:
    AP16349A

 

 

Western Blot: 1:1000

Type: Primary
Antigen: OR4F17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human