Order Entry
United States
ContactUsLinkComponent
Anti-NETO2 Rabbit Polyclonal Antibody
Anti-NETO2 Rabbit Polyclonal Antibody
  89513-016
 :  Abgent
Anti-NETO2 Rabbit Polyclonal Antibody
  89513-016
 :  Abgent
 :  AP13322A

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Neuropilin (Nrp) And Tolloid (Tll)-Like 2
  • Antigen Symbol:
    NETO2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    N-Terminal
  • Gene ID:
    81831
  • Antigen Synonyms:
    BTCL2|NEOT2
  • Amino Acid Number:
    174 to 203
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    59.393 kDa
  • Sequence:
    RSSQVEQEEKTKPGQC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.4mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This NETO2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 174-203 amino acids from the N-terminal region of human NETO2.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89513-016
  • Supplier no.:
    AP13322A

 

 

Western Blot: 1:1000, Immunhistochemsitry: 1:10~50, Flow Cytometry: 1:10~50,

Type: Primary
Antigen: NETO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human