Proliant Biologicals’ Bovine Serum Albumin (BSA), pH 7.0, Standard Grade is a high purity, lyophilized “Fraction V” powder derived from the plasma of USDA-inspected healthy animals from abattoirs located in the USA. This BSA is isolated from other plasma protein products and lipids by a unique proprietary, non-solvent procedure. The separation process includes a modified Cohn/heat-shock procedure designed to inactivate proteases and other potentially interfering enzymes. Typical purity levels are in excess of 99%. It ideal for most diagnostic and research applications.
- High Purity BSA
- Protease Free
- Endotoxin Free
- IgG Free
- Origin Traceability
- Closed Loop Manufacturing
Proliant Biologicals is a manufactuerer of Bovine Serum Albumin who utilizes USDA Edible Grade Plasma and controls the entire process, from collection, through transport and processing.
- CAS No.:9048-46-8
- Source:Serum
- Species:Bovine
- Storage Conditions:Store sealed in a cool, dry environment
- Form:White to yellow with tan to green cast Lyophilized powder
- Endotoxin-free:Yes
- Protease-free:Y
- Protein Synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/Peptide Name:Bovine Serum Albumin
- Purity:100%
- Color:White to yellow with tan to green cast
- Concentration:≥ 98.0%
- pH:6.5–7.5
- Grade:Standard Grade
- Tested Applications:RIA, EIA, Chemiluminescent, Enzyme Stabilizer, etc.