ContactUsLinkComponent
BD BBL™ Taxo™ Differentiation Discs for Haemophilus Species, BD Diagnostic
BD BBL™ Taxo™ Differentiation Discs for Haemophilus Species, BD Diagnostic
Catalog # 90002-550
Supplier:  BD
CAS Number:  
BD BBL™ Taxo™ Differentiation Discs for Haemophilus Species, BD Diagnostic
Catalog # 90002-550
Supplier:  BD
Supplier Number:  231727
CAS Number:  
Specifications
  • Description:
    Differentiation Discs for Haemophilus Species
  • Cat. No.:
    90002-550
  • Supplier no.:
    231727
Specifications
About this item
BD BBL™ Taxo™ Differentiation Discs ideal for differentiating Haemophilus SPP.
  • Ideal for differentiating Haemophilus SPP
  • Based on X or V factors or both
Differentiation is based on their requirements for X or V factors or both.