ContactUsLinkComponent
All in One pH Electrodes, Waterproof Series, Oakton Instruments
All in One pH Electrodes, Waterproof Series, Oakton Instruments
Catalog # 101076-302
undefined
All in One pH Electrodes, Waterproof Series, Oakton Instruments
Catalog # 101076-302
Supplier Number:  WD-35808-72
Specifications
  • Ø:
    12 mm
  • Description:
    All-in-One Waterproof pH electrode, double junction, epoxy body, sealed
  • Cat. No.:
    101076-302
  • Cable length:
    0.91 m (3 ft.)
  • Supplier no.:
    WD-35808-72
Specifications
About this item
All-in-One Waterproof pH electrodes feature a built-in temperature sensor for taking readings with Automatic Temperature Compensation (ATC).
  • Includes a FREE electrode saver bottle to extend electrode life
  • Feature a BNC connector for pH and a waterproof 6-pin connector for temperature
This double junction electrode is suitable for most applications, including dirty water, heavy metals and organics applications.