Alpet D2 Surface Sanitizer/Disinfectant, Hardy Diagnostics
Evaporative isopropyl/quat-based sanitizer Alpet D2 is designed for water-sensitive equipment and low-moisture areas.
- Noncorrosive
- Approved as Boot Sanitizer
- Surface Sanitizer Spray Approved for Food-Contact Surfaces
- Excellent for Packaging Areas, Scales, and Display Cases
A combination of isopropyl alcohol and a powerful quaternary ammonium compound, the sanitizer is effective against norovirus and avian influenza H9N2
It's also used on other items such as freezers, control panels, proximity switches, tools, the inside of electrical panels, motors, thermometers, labelers, utensils, scales, and belts. Treated surfaces must remain wet for five minutes.
It's expected to inactivate all influenza A viruses including Pandemic 2009 H1N1 (swine flu) and eliminate 99% of detectable pathogens.
: NSF certified D2 classification.
Frequently Bought Together