PYREX® Media Storage Bottle with High Temperature Cap, Corning
Supplier: Corning
Certificates
About this item
Heavy duty bottles can be used for storage, mixing, and sampling.
Bottles have permanent white enamel graduations and a marking spot.
Ordering information: Includes red PBT cap and ETFE pouring ring for dry heat sterilization.
Frequently Bought Together