ContactUsLinkComponent
Universal Style Biopsy Punch Handle, OR Grade, Sklar
Universal Style Biopsy Punch Handle, OR Grade, Sklar
Catalog # 10198-894
Supplier:  Sklar
undefined
Universal Style Biopsy Punch Handle, OR Grade, Sklar
Catalog # 10198-894
Supplier:  Sklar
Supplier Number:  90-7560
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Handle Only for Universal Style Biopsy Punch
  • Material:
    Stainless steel
  • Cat. No.:
    10198-894
  • Supplier no.:
    90-7560
Specifications
About this item
Sklar's Universal Style Biopsy Punch Handle is a double-spring pistol grip style handle with a horn on the upper handle, and two finger rests and a slight hook on the lower handle
This device is supplied as a handle only. It may be used with a universal style biopsy punch.