GeneMate Digital Dry Baths, Benchmark Scientific
Supplier: GeneMate
Certificates
About this item
Single, double, and four block capacity.
- Temperature range: Ambient +5 to 150°C
- Temperature accuracy/uniformity: ± 0.2°C
- Temperature increments: 0.1°C
- Timer: 1 to 999 minutes
- Touchpad digital display with built-in timer
- Easy to use block lifter included
- High grade aluminum blocks, sold separately
Frequently Bought Together