ContactUsLinkComponent
Wartenberg Pinwheel, OR Grade, Sklar
Wartenberg Pinwheel, OR Grade, Sklar
  10194-696
 :  Sklar
undefined
Wartenberg Pinwheel, OR Grade, Sklar
  10194-696
 :  Sklar
 :  06-3375
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Wartenberg Pinwheel
  • Environmentally Preferable:
  • Cat. No.:
    10194-696
  • Supplier no.:
    06-3375
This also known as a Wartenberg Neurowheel, is an instrument used to determine nerve sensitivity as it is rolled across the skin.
  • The handle is offset
  • Made of stainless steel
The wheel is comprised of 19 to 21 short, evenly spaced, sharp steel pins.