ContactUsLinkComponent
Spear Point Comedone Extractor, OR Grade, Sklar
Spear Point Comedone Extractor, OR Grade, Sklar
Catalog # 10193-978
Supplier:  Sklar
undefined
Spear Point Comedone Extractor, OR Grade, Sklar
Catalog # 10193-978
Supplier:  Sklar
Supplier Number:  06-4126
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Spear Point Comedone Extractor
  • Material:
    Stainless steel
  • Cat. No.:
    10193-978
  • Supplier no.:
    06-4126
Specifications
About this item
Sklar's Spear Point Comedone Extractor is a dermal instrument used to treat facial blemishes.
  • Double ended
  • Reusable
  • Latex-free
This instrument features one sharp, spear-shaped end for use in rupturing pustules and a round, cup-shaped end with a small round hole specifically designed for extraction. Gentle pressure is applied to the appropriate end when the round opening is placed around the blemish, and the instrument assists in forcing the debris causing the blemish out from under the skin.