ContactUsLinkComponent
Micro Sterilizing Case, Roboz Surgical
Micro Sterilizing Case, Roboz Surgical
Catalog # 100189-710
Supplier:  Roboz Surgical
Micro Sterilizing Case, Roboz Surgical
Catalog # 100189-710
Supplier:  Roboz Surgical
Supplier Number:  RS-9900A
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Dimensions:
    8 x 8.5 x 1.25"
  • Cat. No.:
    100189-710
  • Supplier no.:
    RS-9900A
Specifications
About this item
An Effective Way Of Protecting Delicate Micro Surgical Instruments For Sterilization
Silicone Rubber Inserts Protect Instruments. Includes Removable Cover and a Sentry removable silicone mat.