Order Entry
United States
ContactUsLinkComponent
Heath Curette 4" 2 mm Sharp/Blunt, Roboz Surgical
Heath Curette 4" 2 mm Sharp/Blunt, Roboz Surgical
Catalog # 100188-966
Supplier:  Roboz Surgical
Heath Curette 4" 2 mm Sharp/Blunt, Roboz Surgical
Catalog # 100188-966
Supplier:  Roboz Surgical
Supplier Number:  RS-6354

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Health Curette
  • Length:
    4"
  • Tip dimensions:
    2 mm
  • Cat. No.:
    100188-966
  • Supplier no.:
    RS-6354

Specifications

About this item

One side sharp and one side blunt.