Specifications
- Cat. No.:100188-824
- Supplier no.:RS-6306
Specifications
About this item
Micro Dissecting Trephine; Tungsten Carbide; 1.8 mm Diameter
The head diameter of these trephines is 1.8 mm. The shaft diameter is 2.33 mm. They work with our Ideal micro drill or other handpieces or drills accepting drill bits with 2.33 mm shaft diameter.
These items are designed for animal research only and are not intended for human use.