ContactUsLinkComponent
Forceps, 5" (12.5 cm) Long, Curved With Suture Tying Platform, 0.3 mm Tip, Roboz
Forceps, 5" (12.5 cm) Long, Curved With Suture Tying Platform, 0.3 mm Tip, Roboz
Catalog # 100190-388
Supplier:  Roboz Surgical
Forceps, 5" (12.5 cm) Long, Curved With Suture Tying Platform, 0.3 mm Tip, Roboz
Catalog # 100190-388
Supplier:  Roboz Surgical
Supplier Number:  RS-5263
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Cat. No.:
    100190-388
  • Supplier no.:
    RS-5263
Specifications
About this item
Rhoton Forceps; Micro Suturing With Tying Platform; Curved; 5" Length, 0.3 mm Tip Width.Rhoton forceps can be used in a variety of surgical situations
The feature of this instrument is its long, narrow tip which is helpful in procedures where you need to grasp and manipulate fine, small tissue in deep cavities. The unusual round, textured grip provides great control of the instrument and the tip