Anti-CD158b (KIR2DL2/L3, NKAT2) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: DX27]
APC anti-human CD158b (KIR2DL2/L3, NKAT2) [DX27]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD158b (KIR2DL2/L3, NKAT2)
Clonality: Monoclonal
Clone: DX27
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
- Catalog No:
- 10758-086
- 10758-088
- Antigen Symbol:
- CD158b (KIR2DL2/L3, NKAT2)
- CD158b (KIR2DL2/L3, NKAT2)
- Antigen Synonyms:
- CD158j (KIR2DS2|CD158b1 (KIR2DL2|CD158b2 (KIR2DL3|KIR-NKAT2|p58.3)|p50.2)|p58.2)|KIR2DL2
- CD158j (KIR2DS2|CD158b1 (KIR2DL2|CD158b2 (KIR2DL3|KIR-NKAT2|p58.3)|p50.2)|p58.2)|KIR2DL2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- DX27
- DX27
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3803
- 3803
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together