Anti-CD11c Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 3.9]
Supplier: Biolegend
Certificates
About this item
Brilliant Violet 711™ anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Rhesus;Squirrel Monkey
Specifications
- Catalog No:
- 301629-BL
- 10757-638
- Antigen Symbol:
- CD11c
- CD11c
- Antigen Synonyms:
- aX integrin|Integrin αX subunit|ITGAX|CR4|p150
- aX integrin|Integrin αX subunit|ITGAX|CR4|p150
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 711
- Brilliant Violet® 711
- Clone:
- 3.9
- 3.9
- Reactivity:
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
- Host:
- Mouse
- Mouse
- Gene ID:
- 3687
- 3687
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 711™ and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 711™ and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together