Order Entry
United States
ContactUsLinkComponent
 
Anti-CD11b Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: M1/70]
 :  Biolegend
 
Anti-CD11b Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: M1/70]
 :  Biolegend

 

  • Catalog No:
  • 101241-BL
  • 10756-730
  • Antigen Symbol:
  • CD11b
  • CD11b
  • Antigen Synonyms:
  • Integrin aM|C3biR|Mo1|CR3|Mac-1|Ly-40|αM integrin|ITGAM
  • Integrin aM|C3biR|Mo1|CR3|Mac-1|Ly-40|αM integrin|ITGAM
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 711
  • Brilliant Violet® 711
  • Clone:
  • M1/70
  • M1/70
  • Reactivity:
  • Baboon, Rabbit, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Baboon, Rabbit, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16409, 3684
  • 16409, 3684
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Modification:
  • Integrin aM, alphaM integrin, Mac-1, Mo1, CR3, Ly-40, C3biR
  • Integrin aM, alphaM integrin, Mac-1, Mo1, CR3, Ly-40, C3biR
  • Immunogen:
  • C57BL/10 splenocytes
  • C57BL/10 splenocytes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 711™ and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 711™ and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

Frequently Bought Together