About this item
Brilliant Violet 785™ anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Common Marmoset;Cynomolgus;Rhesus;Swine (Pig;Porcine)
Specifications
- Catalog No:
- 10757-626
- 10757-624
- Antigen Symbol:
- CD11b
- CD11b
- Antigen Synonyms:
- C3biR|Mo1|CR3|Integrin αM chain|Mac-1|ITGAM|αM Integrin
- C3biR|Mo1|CR3|Integrin αM chain|Mac-1|ITGAM|αM Integrin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 785
- Brilliant Violet® 785
- Clone:
- ICRF44
- ICRF44
- Reactivity:
- Baboon, Pig, African green monkey, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human
- Baboon, Pig, African green monkey, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3684
- 3684
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 785™ and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 785™ and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT