Order Entry
United States
ContactUsLinkComponent
Anti-CD11a Mouse Monoclonal Antibody [clone: TS2/4]
Anti-CD11a Mouse Monoclonal Antibody [clone: TS2/4]
  350602-BL
 :  Biolegend
Anti-CD11a Mouse Monoclonal Antibody [clone: TS2/4]
  350602-BL
 :  Biolegend
 :  350602

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD11a
  • Clonality:
    Monoclonal
  • Clone:
    TS2/4
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 μg
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    3683
  • Antigen Synonyms:
    Integrin αL subunit|ITGAL|LFA-1α chain|αL Integrin
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    HLA-DR CTL line
  • Purification:
    The antibody was purified by affinity chromatography.
  • Cat. No.:
    350602-BL
  • Supplier no.:
    350602

 

 

Purified anti-human CD11a [TS2/4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IP; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: TS2/4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human