Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Integrin Alpha L
- Antigen Symbol:CD11A
- Clonality:Monoclonal
- Clone:B-B15
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:200 µg
- Antigen Synonyms:LFA-1 alpha|LFA-1|Leukocyte adhesion glycoprotein LFA-1 alpha chain|antigen CD11A (p180)|LFA1A|Leukocyte function-associated molecule 1 alpha chain|CD11A|alpha polypeptide|CD11 antigen-like family member A|LFA-1A|lymphocyte function-associated antigen 1
- Cat. No.:102995-474
- Supplier no.:CDM027
Type: Primary
Antigen: CD11A
Clonality: Monoclonal
Clone: B-B15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human