Order Entry
United States
ContactUsLinkComponent
Anti-LRRC74B Rabbit Polyclonal Antibody
Anti-LRRC74B Rabbit Polyclonal Antibody
Catalog # 89515-682
Supplier:  Abgent
Anti-LRRC74B Rabbit Polyclonal Antibody
Catalog # 89515-682
Supplier:  Abgent
Supplier Number:  AP19100B

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Leucine Rich Repeat Containing 74B
  • Antigen Symbol:
    LRRC74B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    C-Terminal
  • Gene ID:
    400891
  • Amino Acid Number:
    355 to 383
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    41.621 kDa
  • Sequence:
    RVEYKKELLPVFRSC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This YV012 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 355-383 amino acids from the C-terminal region of human YV012.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89515-682
  • Supplier no.:
    AP19100B

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: LRRC74B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human