Order Entry
United States
ContactUsLinkComponent
Supercut Sklarhone™ Gorney-Freeman Rhytidectomy Scissors, OR Grade, Sklar
Supercut Sklarhone™ Gorney-Freeman Rhytidectomy Scissors, OR Grade, Sklar
Catalog # 10194-186
Supplier:  Sklar
undefined
Supercut Sklarhone™ Gorney-Freeman Rhytidectomy Scissors, OR Grade, Sklar
Catalog # 10194-186
Supplier:  Sklar
Supplier Number:  15-3572

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Supercut Sklarhone™ Gorney-Freeman Rhytidectomy Scissors
  • Length:
    22.9 cm (9")
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. No.:
    10194-186
  • Supplier no.:
    15-3572
  • Blade type:
    Blunt/Blunt

Specifications

About this item

Supercut Sklarhone™ Gorney-Freeman Rhytidectomy (face lift) Scissors are constructed of stainless steel

They feature one micro-serrated edge and one razor edge, with semi-sharp outer edges and flat, tapered tips. Industry standard.