Stainless Steel Syringe Filter Holders, Advantec MFS
Supplier: Advantec MFS
Certificates
About this item
These stainless-steel syringe holders are economical and re-usable thereby reducing waste.
- PTFE O-ring and gasket
- May be autoclaved with filter in place
- Tightened securely with included wrench set
Applications include cleaning small volumes of liquid such as eyewash or photoresist, clear turbid solutions such as serums or purify virus solutions. This also can be used in preventing leakage from or contamination of vented apparatus such as vacuum manifolds or vacuum pumps.
Specifications
- Catalog No:
- 101104-832
- 101105-078
- Inlet/Outlet Connections:
- Female Luer-Lok/Male Luer Slip
- Female Luer-Lok/Male Luer Slip
- Max. Differential Pressure:
- 42 psi
- 42 psi
- Max. Inlet Pressure:
- 98 psi
- 98 psi