Order Entry
United States
ContactUsLinkComponent
 
Urine Albumin-to-Creatinine Ratio Uacr as Kit, BioVision
Catalog # 10013-754
Supplier:  Avantor
CAS Number:  
undefined
Urine Albumin-to-Creatinine Ratio Uacr as Kit, BioVision
Catalog # 10013-754
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:
  • Storage Conditions:
    -20°C
  • Species Reactivity:
    Mammalian samples
  • Sample Type:
    Albumin: urine, saliva, etc.; Creatinine: urine, serum, etc.; UACR: urine
  • Shelf Life:
    12 Months
  • Cat. No.:
    10013-754
  • No. of assays/kit:
    100 assays
  • Supplier no.:
    K551-100

Specifications

About this item

Only kit on the market to measure UACR

  • Detection method- Fluorescence (Ex/Em = 600/630 nm) & Absorbance (OD 570 nm)
  • Applications- Estimation of albumin in biological samples
  • - Estimation of creatinine in biological samples
  • - Determination of UACR in mammalian urine samples

Urine Albumin-to-Creatinine Ratio (UACR) is one of the two markers used to determine chronic kidney disease (CKD). UACR is recommended to be measured on regular basis on people living with Type I and Type II diabetes. UACR is defined as the ratio between albumin (reported in mg/dl) and creatinine (reported in g/dl). This ratio estimates the amount of albumin excreted in urine during a 24 hr period. Albuminuria is diagnosed when UACR is greater than 30 mg albumin/g creatinine. BioVision’s UACR Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects albumin (detection range: 0.02- 2.5 mg/ml), creatinine (detection range: 0.002 -0.5 mg/ml) and urine albumin-to-creatinine ratio. The UACR ratio is determined in two steps: First, albumin is determined by using a probe (AB580) that specifically recognizes albumin (Ex/Em = 600/630 nm). Second, creatinine is converted to sarcosine via enzymatic reactions. Sarcosine is specifically oxidized generating a product that reacts with a probe producing a chromophore that can be detected at 570 nm.