Specifications
- Description:Granulated Activated Copper, 30 mesh
- Cat. No.:10803-394
- Type:SPE Accessory
- Supplier no.:26136
Specifications
About this item
Activated copper effectively removes elemental sulfur from environmental extracts.
- Convenient form for removing sulfur from environmental extracts
- Acidified and activated—ready for use
Restek's acid-washed and activated material can be used right out of the package. The 30 mesh granular material eliminates the potential for fine copper particles in filtered extracts.