Resprep® SPE Cartridges (Normal Phase), Restek
Supplier: Avantor
Certificates
About this item
Resprep® SPE cartridges contain hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
- Column hardware constructed of glass with PTFE frits or only PTFE
- Volume varies between models
Compatible with EPA SW 846 methods and CLP protocols, depending on models.
Packaging: May be packed with either florisil or silica.
Frequently Bought Together