- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Conjugation:Cy5®
- Host:Goat
- Reactivity:Mouse
- Size:150 µg
- Cat. No.:95040-046
- Supplier no.:PA45009
ECL Plex Cy2-, Cy3-, and Cy5-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent Western blotting.
- For 1000 cm² membrane area
ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras. Products listed below are optimized for use with ECL Plex Western Blotting System:
• ECL Plex CyDye-conjugated antibodies: A43009, PA43010, PA45009, PA45010, PA45011, PA45012
• Hybond ECL: RPN2020D, RPN78D, RPN203D, and RPN3032D
• Hybond-LFP: RPN2020LFP, RPN2020LFP3, RPN1416LFP, and RPN303LFP
• ECL Plex fluorescent rainbow markers: RPN850 and RPN85
• Blocking buffer: RPN412
• ECL Plex combination packs: RPN998 and RPN999
ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse