About this item
>98% Pure active Recombinant human BMP-4.
BioVision’s recombinant human BMP-4 is biologically active. The ED₅₀ as determined by dose-dependent induction of alkaline phosphatase activity with C2C12 cells is in the range of 2-12 µg/ml.
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Recombinant human BMP-4 produced from E. coli is a 13 kDa monomeric, non-glycosylated, polypeptide chain containing 116 amino acid residues.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Endotoxin Content:<0.1 ng/μg
- Reconstitution Instructions:Reconstitute to a concentration of 0.1-1.0 mg/ml in sterile 20 mM acetic acid.
- Protein Synonyms:BMP-4|rh-BMP-4|BMP4|BMP 4|h-BMP-4
- Protein/Peptide Name:BMP4
- Purity:>98%
- Molecular Weight:13.0 kDa
- Formulation:Sterile filtered and lyophilized powder