Order Entry
United States
ContactUsLinkComponent
Human Recombinant Long R3 Igf-I (from E. coli)
Human Recombinant Long R3 Igf-I (from E. coli)
Supplier:  Avantor
Certificates
undefined
Human Recombinant Long R3 Igf-I (from E. coli)
Supplier:  Avantor

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Endotoxin Content:
    <0.1 ng/μg
  • Reconstitution Instructions:
    Reconstitute in dH₂O or 10 mM AcOH to 1.0 mg/ml.
  • Protein Synonyms:
    Insulin-like Growth Factor-1|IGF-IA|Somatamedin C
  • Protein/Peptide Name:
    Long R3 Igf-I
  • Purity:
    >97%
  • Molecular Weight:
    9.11 kDa
  • Formulation:
    Lyophilized with no additives

Specifications

Related Information

Frequently Bought Together