Mouse Recombinant PDGF-BB (from E. coli)
Supplier: Avantor
Certificates
About this item
>95% Pure active Recombinant murine PDGF-BB .
The ED₅₀ as determined by the dose-dependent proliferation of murine BALB/c 3T3 cells is < 2 ng/ml.
Recombinant mouse Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle and connective tissue. PDGF, which is composed of a homodimeric, non-glycosylated, polypeptide of 109 amino acids and having a molecular mass of 24.4 kDa.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Endotoxin Content:<0.1 ng/μg
- Reconstitution Instructions:Reconstitute in sterile 100mM acetic acid and 0.1% BSA to a concentration of 0.1-1.0 mg/ml.
- Protein Synonyms:PDGF-BB|m-PDGF-BB|rm-PDGF-BB|PDGF BB|PDGFBB
- Protein/Peptide Name:PDGF-BB
- Purity:>95%
- Molecular Weight:24.4 kDa
- Formulation:Lyophilized with no additives
Specifications
Frequently Bought Together