Nitrile Exam Gloves, Powder-Free Fingertip-Textured, Tronex®
Supplier: Tronex
Certificates
About this item
Advanced nitrile material formulation provides superior tactile sensitivity with lightness of fit, reduced hand fatigue, and excellent barrier protection from bloodborne pathogens
- 100% latex-free, powder-free, and nonsterile
- Fingertip textured for improved grip
- Outstanding comfort, durability, and elasticity
- Ambidextrous with beaded cuffs
Offers exceptional resistance to chemicals, oils, and temperature extremes when compared to latex or vinyl gloves.
Specifications
- Catalog No:
- 10818-554
- 10818-552
- 10818-556
- 10818-568
- 10818-566
- 10818-570
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- ASTM No.:
- D6319, F1671
- D6319, F1671
- D6319, F1671
- D6319, F1671
- D6319, F1671
- D6319, F1671