Anti-beta Tubulin Mouse Monoclonal Antibody [clone: BT7R]
Catalog # 102963-600
Supplier: Cell Biolabs
Specifications
- Antibody Type:Primary
- Antigen Name:beta Tubulin
- Clonality:Monoclonal
- Clone:BT7R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rabbit,Monkey,Chicken,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Format:Purified
- Form:Liquid
- Storage Temperature:–20 °C
- Shipping Temperature:4°C
- Cat. No.:102963-600
- Supplier no.:AKR-009
Specifications
About this item
Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: BT7R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Rabbit, Chicken, Monkey