Multilingual Wide Mouth Wash Bottles
Catalog # 490017-202
Supplier: Dynalon
Specifications
- Capacity:500 ml
- Description:Combo Pack (1 of each)
- Cat. No.:490017-202
- Supplier no.:506495-0005
Specifications
About this item
Multi-lingual wide mouth wash bottles feature a patented DripLok® vapor venting valve that prevents solvent drips.
- Indelibly printed in four languages: English, French, German, and Spanish
- Closure features precision jet-tip aperture for fine control.
Choose from four solvent labels, all including the new Globally Harmonized Symbols, NFPA diamond, risk phrases, and safety information.