Order Entry
United States
ContactUsLinkComponent
Lab-Tip® 125S Cleanroom Swab, Berkshire
Lab-Tip® 125S Cleanroom Swab, Berkshire
Catalog # 89496-568
Supplier:  Berkshire
Small Business Enterprise
undefined
Lab-Tip® 125S Cleanroom Swab, Berkshire
Catalog # 89496-568
Supplier:  Berkshire
Supplier Number:  LTP125S.25
Small Business Enterprise

Specifications

  • Description:
    BCR® Large Polyester Knit Sampling Swab
  • Cat. No.:
    89496-568
  • Overall length:
    125 mm (4.92")
  • Handle material:
    Polypropylene
  • Head type:
    Large semi-flexible paddle
  • Supplier no.:
    LTP125S.25

Specifications

About this item

The Lab-Tip® 125S Large Sampling Swab is a cleanroom-laundered, double-layered polyester knit swab.

  • Autoclavable
  • Low particles, fibers, ions, and extractables
  • Excellent chemical resistance
  • Soft and non-abrasive with no contaminating adhesives
  • Good sorbency
  • Free from silicone, amides, and DOP
  • Cleanroom Class 100 to 100000

Grooves on the handle make for easy snapping. The cleanroom-laundered knitted head provides ultra-low NVR and ion levels. Contamination control, with critically low particles, fibers, ions, and extractables. Excellent chemical resistance. Soft and non-abrasive.

It is designed for general cleaning surface sampling in cleanrooms and controlled environments in many industries.

Ordering information: 20 swabs per pack, 25 packs per case (double pack).