Half Circle Fistula Needle, OR Grade, Sklar
Supplier: Sklar
Certificates
About this item
Sklar's Half Circle Stainless Steel Fistula Needle is used in veterinary suturing procedures.
The half circle is useful in confined surgical sites, although it requires the user to move his wrist more.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.