Human Recombinant BDNF (from E. coli)
Supplier: Avantor
Certificates
About this item
A potent neurotrophic factor supporting the growth and survival of nerve/glial cells
The ED₅₀ determined by its ability to stimulate chick E8 neurite outgrowth was found to be 0.1-1.0 ng/ml.
Human Brain-Derived Neurotrophic Factor (BDGF) is a potent neurotrophic factor that supports the growth and survivability of nerve and glial cells. The active form of BDNF is a 27.0 kDa dimer, formed by two identical 119 amino acid subunits held together by strong hydrophobic interactions.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Endotoxin Content:<0.1 ng/μg
- Reconstitution Instructions:Reconstitute in H₂O to a concentration of 0.1-1.0 mg/ml.
- Protein Synonyms:h-BDNF|BDNF|rh-BDNF
- Protein/Peptide Name:BDNF
- Purity:>98%
- Molecular Weight:27.0 kDa
- Formulation:Sterile filtered and lyophilized with no additives
Specifications
Frequently Bought Together