ContactUsLinkComponent
Kevorkian Micro-Cut Biopsy Punch Canula and Tip, OR Grade, Sklar
Kevorkian Micro-Cut Biopsy Punch Canula and Tip, OR Grade, Sklar
Catalog # 10184-204
Supplier:  Sklar
undefined
Kevorkian Micro-Cut Biopsy Punch Canula and Tip, OR Grade, Sklar
Catalog # 10184-204
Supplier:  Sklar
Supplier Number:  90-8976
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Kevorkian Micro-Cut Biopsy Punch Canula
  • Material:
    Stainless steel
  • Tip configuration:
    Rectangular Bite with 4 Lower Teeth
  • Cat. No.:
    10184-204
  • Shape:
    Rectangular
  • Supplier no.:
    90-8976
  • Tip type:
    Straight
Specifications
About this item
Sklar's Kevorkian Micro-Cut Biopsy Punch Canula and Tip is part of an OB/GYN instrument
This product attaches to a universal style handle to create biopsy punch forceps. They are used in colposcopy procedures to obtain cervical tissue samples for biopsy. The bite of this Kevorkian punch is 3mm x 8mm and it produces an rectangular shaped tissue sample. It features four lower teeth, useful for grasping and holding difficult to reach surface tissue; however, are not optimal when a deep tissue sample is desired. The length of this product, which is the cannula and the tip only, is approximately 8 inches.