Conductive Dividable Grid Containers, Quantum Storage Systems
Supplier: QUANTUM STORAGE SYSTEMS
Certificates
About this item
Protect your electronic components from typical electronmagnetic interference, electrostatic or induced fields with molded carbon-filled, copolymer polypropylene bins.
Conductive material provides a barrier which these fields cannot penetrate and prevents the build-up of electrostatic charge. Bins are permanently conductive and are unaffected by washing with normal cleaners.
Weigh capacity of 100 lbs. Divider available to customize your container into compartments at 11/8" Intervals.
Surface resistivity is less than 105 Ohms-per square. Black color.
Ordering information: Items must be ordered in increments of 12.
Frequently Bought Together