Order Entry
United States
ContactUsLinkComponent
Mouse Monoclonal IgG1 Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Mouse Monoclonal IgG1 Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 10820-956
Supplier:  Tonbo Biosciences
undefined
Mouse Monoclonal IgG1 Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 10820-956
Supplier:  Tonbo Biosciences
Supplier Number:  65-4714-U100

Specifications

  • Antibody Type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Storage Temperature:
    Store undiluted at 2-8°C, protect from light, do not freeze.
  • Cat. No.:
    10820-956

Specifications

About this item

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: Unknown specificity
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PerCP-Cy®5.5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: