Rat Monoclonal IgG2a Isotype Control [clone: 2A3]
10820-958
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Size:100 µg
- Cross Adsorption:No
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Cat. No.:10820-958
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.
Type: Control
Antigen: Unknown specificity
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: