About this item
The VWR® Plus Series Solid/Glass Door Compact Laboratory Refirgerators feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
- Temperature Controller
- Digital setting accuracy
- Precise temperature maintenance
- Monitoring and alarms
- Digital temperature display
- High/low temperature alarms
- Remote alarm contacts
Uniformed forced air directional cooling
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
The all new digital temperature display and alarm module offers industry leading product security with a monitoring and alarm system that is 100% independent of the temperature controller. Real time sample simulated temperatures are constantly displayed and supported by audible/visual high and low temperature alarms, along with power outage and sensor error alarms. Standard features include: Keyed door locks, probe access port.