ContactUsLinkComponent
Mini to Standard USB Adapter
Mini to Standard USB Adapter
Catalog # 470219-302
undefined
Mini to Standard USB Adapter
Catalog # 470219-302
Supplier Number:  MINI-USB
Specifications
  • Description:
    Mini to standard USB adapter
  • Cat. No.:
    470219-302
  • Supplier no.:
    MINI-USB
Specifications
About this item
This adapter allows an EasyTemp or EasyLink to be connected to a computer for data collection using Logger Pro or Logger Lite software. The adapter has a USB Mini-A receptacle that connects to the EasyTemp or EasyLink and a standard-A plug that connects to the computer. It can also be used with the USB cable that comes with the CBR 2.
Connector 1: USB Mini Type A Receptacle
Connector 2: USB Standard Type A Plug
Note: Most consumer electronic devices use a USB Mini Type B plug, and will NOT WORK with this adapter. Our adapter only works for devices with USB Mini Type A plug.