ContactUsLinkComponent
Vernier® AC Adapter
Vernier® AC Adapter
  470219-286
undefined
Vernier® AC Adapter
  470219-286
 :  IPS
  • Description:
    Vernier AC Adapter
  • Cat. No.:
    470219-286
  • Supplier no.:
    IPS
This is a 6-volt, DC, tip-negative power supply for use with Vernier LabPro, CBL 2, Digital Control Unit, and Wireless Dynamics Sensor System.