ContactUsLinkComponent
Analog Breadboard Cable
Analog Breadboard Cable
Catalog # 470219-232
undefined
Analog Breadboard Cable
Catalog # 470219-232
Supplier Number:  BB-BTA
Specifications
  • Description:
    Analog Breadboard Cable
  • Cat. No.:
    470219-232
  • Supplier no.:
    BB-BTA
Specifications
About this item
The Breadboard Cable (Analog) provides an easy way for students to build their own sensor circuitry and input the signal into a Vernier interface for data acquisition.
One end of the cable is a standard British Telecom Analog (BTA) connector that plugs into Vernier interfaces. The other end of the cable is a prototyping board connector that exposes the six sensor lines on a prototyping board.
Pin Information: There are six pins available: SIG1, 5V, ID, VRES, GND, and SIG2.