Order Entry
United States
Orders LinkContactUsLinkComponent
Cycloheximide 90%

Cycloheximide 90%

Toxic
Health
Environment
Cycloheximide 90%
Certificates
Cycloheximide 90%
Cycloheximide 90%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis.

Purity: 90% CAS No.: 66-81-9 Molecular Formula: C15H23NO4 Formula Weight: 281.35 Flash Point: 110°(230°F) Physical Form: Powder Melting Point: 111-116° Boiling Point: 492° Solubility: Soluble in water at 20mg/ml Beilstein Reference: 88868 Merck Reference: 14,2728 UN No.: 2811 MDL No.: MFCD00082346

A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis.

Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35
  • Merck Index:14,2728
  • Boiling Pt:492 °C
  • Melting Pt:111…116 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,III

Documents

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $70.93
    $70.93
    Each (1g)
       $70.93

    Supplier Number: J66665-03
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $226.68
    $226.68
    Each (5g)
       $226.68

    Supplier Number: J66665-06
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.