Order Entry
United States
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution solution, standard sensitivity ready-to-use

3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution solution, standard sensitivity ready-to-use

Flammable
Toxic
Health
3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution solution, standard sensitivity ready-to-use
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution solution, standard sensitivity ready-to-use

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.

CAS No.: 54827-17-7 Molecular Formula: C16H20N2 Formula Weight: 240.35 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pale yellow Beilstein Reference: 2808541 MDL No.: MFCD00007748

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:259-364-6
  • UN:1992
  • ADR:3,III

Documents

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 125 mL
    Log in for availability
    Log in for availability
    Each (125ml)
    Each (125ml) - $184.67
    $184.67
    Each (125ml)
       $184.67

    Supplier Number: J63202-EQE
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $276.92
    $276.92
    Each (500ml)
       $276.92

    Supplier Number: J63202-AP
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 1000 mL
    Log in for availability
    Log in for availability
    Each (1ml)
    Each (1ml) - $420.64
    $420.64
    Each (1ml)
       $420.64

    Supplier Number: J63202-AU
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.