Order Entry
United States
ContactUsLinkComponent
Cerium, sputtering target, Diameter 76.2 mm, Thickness 3,18 mm
Flammable
Cerium, sputtering target, Diameter 76.2 mm, Thickness 3,18 mm
Catalog # AA45926-KS
CAS Number:  
Flammable
Cerium, sputtering target, Diameter 76.2 mm, Thickness 3,18 mm
Catalog # AA45926-KS
Supplier Number:  045926-KS
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 each
  • Cat. No.:
    AA45926-KS
  • Supplier no.:
    45926-KS

Specifications

About this item

CAS Number:7440-45-1
 :
Formula:Ce
MDL Number:MFCD00010924
 :140.12 g/mol
Storage Temperature:Ambient
EINECS:231-154-9
UN:1333
ADR:4.1,II
 :GHS02