Fused Quartz Microscope Slides, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Jump to:
About this item
These quartz discs are manufactured by fusing naturally occuring crystalline silica.
- High chemical and corrosion resistance
- Ground and polished
- High melting point
- Extreme hardness
- Low coeffiecent of thermal expansion
- Excellent electrical insulation quality
- Optical transmission from ultra-violet to infra-red
6 Options Available Below
Product Details & Documents
These quartz discs are manufactured by fusing naturally occuring crystalline silica.
- High chemical and corrosion resistance
- Ground and polished
- High melting point
- Extreme hardness
- Low coeffiecent of thermal expansion
- Excellent electrical insulation quality
- Optical transmission from ultra-violet to infra-red
Documents
Product Family Options
Product Information
- ThicknessL×WAvailabilityPrice
- AA42295-KGQuartz slides FU 1 PCSupplier Number: 042295-KG
- AA42295-KJQuartz slides FU 5 PCSupplier Number: 42295-KJ
- AA42296-KGQuartz slides FU 1 PCSupplier Number: 042296-KG
- AA42296-KJQuartz slides FU 5 PCSupplier Number: 042296-KJ
- AA42297-KGQuartz slides FU 1 PCSupplier Number: 042297-KG
- AA42297-KJQuartz slides FU 5 PCSupplier Number: 042297-KJ