

Graphite (natural) ≥99.9995% (metals basis), powder -200 mesh, briquetting grade
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7782-42-5
:
Formula:C
MDL Number:MFCD00144065
ADR:4.1,III
:12.01 g/mol
EINECS:231-955-3
UN:1325
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberAlfa Aesar Metals and Materials
Product Literature
Alfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Frequently Bought Together